CAS RN 100-26-5
CAS RN 100-26-5
IUPAC Name:
pyridine-2,5-dicarboxylic acid
Sinónimos:
2,5-pyridinedicarboxylic acid isocinchomeronic acid
Peso molecular (g/mol):
167.12
Fórmula molecular:
C7H5NO4
InChi Key:
LVPMIMZXDYBCDF-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(N=C1)C(O)=O