CAS RN 10034-98-7
CAS RN 10034-98-7
IUPAC Name:
diytterbium(3+) octahydrate trisulfate
Sinónimos:
diytterbium(3+) octahydrate trisulfate
Peso molecular (g/mol):
778.4
Fórmula molecular:
H16O20S3Yb2
InChi Key:
MDOSIOHMOWGVIM-UHFFFAOYSA-H
SMILES:
O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
1
–
1
de
1
resultados
1
Sulfato de iterbio(III) octahidrato, REacton™, 99,9 % (REO), Thermo Scientific Chemicals
CAS: 10034-98-7 Fórmula molecular: H16O20S3Yb2 Peso molecular (g/mol): 778.40 Número MDL: MFCD00149887 Clave InChI: MDOSIOHMOWGVIM-UHFFFAOYSA-H Sinónimo: unii-e5w6do92vs,e5w6do92vs,3so4.2yb.8h2o,ytterbium sulfate,,ytterbium sulfate octahydrate,ytterbium sulfate,ytterbium iii sulfate octahydrate, reacton,diytterbium 3+ octahydrate trisulfate,sulfuric acid,ytterbium 3+ salt 3:2 , octahydrate 8ci,9ci PubChem CID: 25022290 SMILES: O.O.O.O.O.O.O.O.[Yb+3].[Yb+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O