CAS RN 1015691-13-0
CAS RN 1015691-13-0
IUPAC Name:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Hidrato de cloruro de tetraetilamonio, 99 %, Thermo Scientific Chemicals
CAS: 1015691-13-0 Fórmula molecular: C8H22ClNO Peso molecular (g/mol): 183.72 Número MDL: MFCD00149992 Clave InChI: CALLTGJPWMIDPC-UHFFFAOYSA-M Sinónimo: tetraethylammonium chloride,tetraethyl ammonium chloride,etamon chloride,tea chloride,teac,n,n,n-triethylethanaminium chloride,tetraethylammoniumchloride,ethanaminium, n,n,n-triethyl-, chloride,tetraethyl-ammonium chloride,unii-8b82z83xnn PubChem CID: 5946 ChEBI: CHEBI:78161 SMILES: O.[Cl-].CC[N+](CC)(CC)CC