CAS RN 1073371-99-9
CAS RN 1073371-99-9
IUPAC Name:
methyl 2-hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Sinónimos:
methyl 2-hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
Peso molecular (g/mol):
278.11
Fórmula molecular:
C14H19BO5
InChi Key:
SRYGMLVCVMGUTB-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(O)C=C(C=C1)B1OC(C)(C)C(C)(C)O1