CAS RN 107792-63-2
CAS RN 107792-63-2
IUPAC Name:
λ⁴-cerium(4+) tetratrifluoromethanesulfonate
Sinónimos:
λ⁴-cerium(4+) tetratriflate
Peso molecular (g/mol):
736.37
Fórmula molecular:
C4CeF12O12S4
InChi Key:
BYAJTISDHIASIP-UHFFFAOYSA-J
SMILES:
[Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
1
–
1
de
1
resultados
1
Trifluorometansulfonato de cerio(IV), 98 %, Thermo Scientific Chemicals
CAS: 107792-63-2 Fórmula molecular: C4CeF12O12S4 Peso molecular (g/mol): 736.37 Número MDL: MFCD03844746 Clave InChI: BYAJTISDHIASIP-UHFFFAOYSA-J Sinónimo: cerium iv trifluoromethanesulfonate,cerium iv triflate,cerium trifluoromethanesulfonate,acmc-20alhh,ceric triflate,cerium iv trifluoromethanesulphonate,cerium iv trifluoromethanesulfonate, ?ce cf3so3 4,trifluoromethanesulfonic acid, cerium iv salt,methanesulfonic acid,1,1,1-trifluoro-,cerium 4+ salt 4:1 PubChem CID: 9875257 Nombre IUPAC: cerio(4+); trifluorometanosulfonato SMILES: [Ce+4].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F