missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C3a Antibody (6E8), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-17998CL1
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Complement C3a Monoclonal antibody specifically detects Complement C3a in Human samples. It is validated for ELISA,Western Blot
Especificaciones
| Complement C3a | |
| Monoclonal | |
| CoraFluor 1 | |
| Acylation Stimulating Protein, acylation-stimulating protein cleavage product, AHUS5, ARMD9, ASP, C3, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, C3a, C3a anaphylatoxin, C3adesArg, C3b, C3bc, C3-beta-c, complement C3, Complement C3 alpha chain, Complement C3 beta chain, Complement C3b alpha' chain, Complement C3c alpha' chain fragment 1, Complement C3c alpha' chain fragment 2, Complement C3d fragment, Complement C3dg fragment, Complement C3f fragment, Complement C3g fragment, complement component 3, complement component C3, complement component C3a, complement component C3b, CPAMD1, EC 3.4.21.43, epididymis secretory sperm binding protein Li 62p, HEL-S-62p, prepro-C3 | |
| Recombinant netrin domain of human C3. UniProt: P01024 | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Autoimmune Diseases, Diabetes Research, Immunology, Inflammation, Innate Immunity, Neuroscience, Signal Transduction, Stem Cells | |
| 718 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| ELISA, Western Blot | |
| 6E8 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido