Learn More
Endecotts™ 100 mm Diameter Stainless Steel Frame and Wire Mesh Test Sieves
Click to view available options
Tamaño de poro:
32 μm
53 μm
1 mm
1,1 mm
125 μm
2 mm
250 μm
2,8 mm
355 μm
425 μm
4,75 mm
600 μm
9,5 mm
Tamaño de la unidad:
cada uno
Descripción
- Made with only the highest quality materials, available in diameter sizes of 38, 100, 150, 200, 250, 300, 315, 350, 400 or 450 mm as well as in 3, 8, 12 or 18 inches.
- Precision frame (ensures consistent nestability)
- Precise aperture (in accordance with ISO 3310-1, ASTM E11 or other specifications)
- Natural fillet (free flowing of sample)
- Totally sealed (no crevice to lose material)
- Evenly tensioned mesh (ensures accurate analysis)
- Safe edge (big radius makes it comfortable to handle)
- Serial number (ensures full traceability)
Especificaciones
Especificaciones
| Cantidad | 1Each |
| Tamaño de poro | 1,1 mm |
| Tipo | Test Sieve |
| Certificaciones/conformidad | ISO 3310-1 & 2 requirements, ASTM, BSI |
| Material | Stainless Steel |
| Para utilizar con (aplicación) | For all types of laboratory sampling and particle size analysis |
| Diámetro exterior (imperial) | 100 mm |
| Material del bastidor | Acero inoxidable |
| Mesh Material | Acero inoxidable |