Learn More
Ácido 2,2-difluorociclopropanocarboxílico, 95 %, Thermo Scientific Chemicals
Catalog Number 11309969
Click to view available options
Cantidad:
250 mg
1 g
5 g
107873-03-0
C4H4F2O2
122.071
MFCD04115828
KMLMOVWSQPHQME-UHFFFAOYSA-N
2,2-difluorocyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 2,2-difluoro, acmc-20aht4, 2-carboxy-1,1-difluorocyclopropane, 2,2-difluoro cyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic?acid, 2,2-difluorocyclopropane carboxylic acid, 2,2-difluorocyclopropane-carboxylic acid, 2,2-difluoro-cyclopropane-carboxylic acid
2782823
ácido 2,2-difluorociclopropano-1-carboxílico
C1C(C1(F)F)C(=O)O
107873-03-0
C4H4F2O2
122.071
MFCD04115828
KMLMOVWSQPHQME-UHFFFAOYSA-N
2,2-difluorocyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic acid, cyclopropanecarboxylic acid, 2,2-difluoro, acmc-20aht4, 2-carboxy-1,1-difluorocyclopropane, 2,2-difluoro cyclopropanecarboxylic acid, 2,2-difluoro-cyclopropanecarboxylic?acid, 2,2-difluorocyclopropane carboxylic acid, 2,2-difluorocyclopropane-carboxylic acid, 2,2-difluoro-cyclopropane-carboxylic acid
2782823
ácido 2,2-difluorociclopropano-1-carboxílico
C1C(C1(F)F)C(=O)O
2,2-Difluorocyclopropanecarboxylic acid derivative (i.e. 2,2-difluorocyclopropanecarbonyl chloride) is used in Friedel-Crafts reactions with various arenes to prepare aryl 2,2-difluorocyclopropyl ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Aplicaciones
El derivado del ácido 2,2-difluorociclopropanocarboxílico (es decir, cloruro de 2,2-difluorociclopropanocarbonilo) se utiliza en las reacciones de Friedel-Crafts con varios arenos para preparar aril 2,2-difluorociclopropil cetonas.
Solubilidad
Soluble en agua.
Notas
Incompatible con agentes oxidantes fuertes.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 2,2-Difluorocyclopropanecarboxylic acid
Punto de fusión 55°C to 60°C
Punto de ebullición 70°C (7 mmHg)
Cantidad 250 mg
Número UN UN3261
Información de solubilidad Soluble in water.
Formula Weight (peso de la fórmula) 122.07
Porcentaje de pureza 95%