missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ácido 2,4-dimetilfenilacético, 98 %, Thermo Scientific™
63.50€ - 220.00€
Identificadores de productos químicos
CAS | 6331-04-0 |
---|---|
Fórmula molecular | C10H12O2 |
Peso molecular (g/mol) | 164.20 |
Número MDL | MFCD02664685 |
Clave InChI | MWBXCWLRASBZFB-UHFFFAOYSA-N |
Sinónimo | 2,4-dimethylphenylacetic acid, 2-2,4-dimethylphenyl acetic acid, 2,4-dimethylphenyl acetic acid, 2,4-dimethyphenylacetic acid, acetic acid, 2,4-xylyl, 2,4-dimethylphenylaceticacid, acetic acid,4-xylyl, ksc495a3d, 2,4-dimethylphenyl-acetic acid, 2,4-dimethyl phenyl acetic acid |
PubChem CID | 138729 |
Nombre IUPAC | Ácido 2-(2,4-dimetilfenil)acético |
SMILES | CC1=CC=C(CC(O)=O)C(C)=C1 |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
6331-04-0 | |
164.20 | |
MWBXCWLRASBZFB-UHFFFAOYSA-N | |
138729 | |
CC1=CC=C(CC(O)=O)C(C)=C1 |
C10H12O2 | |
MFCD02664685 | |
2,4-dimethylphenylacetic acid, 2-2,4-dimethylphenyl acetic acid, 2,4-dimethylphenyl acetic acid, 2,4-dimethyphenylacetic acid, acetic acid, 2,4-xylyl, 2,4-dimethylphenylaceticacid, acetic acid,4-xylyl, ksc495a3d, 2,4-dimethylphenyl-acetic acid, 2,4-dimethyl phenyl acetic acid | |
Ácido 2-(2,4-dimetilfenil)acético |
Especificaciones
2,4-Dimethylphenylacetic acid | |
C10H12O2 | |
2,4-dimethylphenylacetic acid, 2-2,4-dimethylphenyl acetic acid, 2,4-dimethylphenyl acetic acid, 2,4-dimethyphenylacetic acid, acetic acid, 2,4-xylyl, 2,4-dimethylphenylaceticacid, acetic acid,4-xylyl, ksc495a3d, 2,4-dimethylphenyl-acetic acid, 2,4-dimethyl phenyl acetic acid | |
CC1=CC=C(CC(O)=O)C(C)=C1 | |
164.20 | |
164.21 |
6331-04-0 | |
MFCD02664685 | |
MWBXCWLRASBZFB-UHFFFAOYSA-N | |
Ácido 2-(2,4-dimetilfenil)acético | |
138729 | |
98% |
Seguridad y manipulación
Declaración de GHS H
H315-H319-H335
Provoca irritación cutánea.
Provoca irritación ocular grave.
Puede irritar las vías respiratorias.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335.
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes