missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Éster de pinacol de ácido 2-(etoxicarbonil)furano-5-borónico, 96 %, Thermo Scientific Chemicals
Click to view available options
Cantidad:
250 mg
1 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Nombre del producto químico o material | 2-(Ethoxycarbonyl)furan-5-boronic acid pinacol ester |
Cantidad | 250 mg |
Sinónimo | ethyl 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-ethoxycarbonyl furan-2-boronic acid, pinacol ester, 5-ethoxycarbonyl furan-2-boronic acid pinacol ester, 2-ethoxycarbonyl furan-5-boronic acid pinacol ester, ethyl 5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, ethyl5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furan-2-carboxylate, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-furan-2-carboxylic acid ethyl ester |
Formula Weight (peso de la fórmula) | 266.1 |
Porcentaje de pureza | 96% |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'