missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopiradicina, 95 %, Acros Organics™
69.42€ - 209.00€
Identificadores de productos químicos
CAS | 5469-70-5 |
---|---|
Fórmula molecular | C4H5N3 |
Peso molecular (g/mol) | 95.1 |
Clave InChI | LETVJWLLIMJADE-UHFFFAOYSA-N |
Sinónimo | 3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin |
PubChem CID | 230373 |
Nombre IUPAC | piridazina-3-amina |
SMILES | C1=CC(=NN=C1)N |
Descripción
Identificadores de productos químicos
5469-70-5 | |
95.1 | |
3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin | |
piridazina-3-amina |
C4H5N3 | |
LETVJWLLIMJADE-UHFFFAOYSA-N | |
230373 | |
C1=CC(=NN=C1)N |
Especificaciones
3-Aminopyridazine | |
168.0°C to 172.0°C | |
94% min. (HPLC) | |
3-aminopyridazine, 3-pyridazinamine, aminopyridazine, pyridazin-3-ylamine, 3-amino-1,2-diazine, pyridazine-3-ylamine, pyridazineamine, pyridazyl amine, imino-pyridazine, 3-pyridazinamin | |
LETVJWLLIMJADE-UHFFFAOYSA-N | |
piridazina-3-amina | |
230373 | |
95% |
5469-70-5 | |
Authentic | |
C4H5N3 | |
Solubility in water: .. Other solubilities: soluble in methanol | |
C1=CC(=NN=C1)N | |
95.1 | |
95.1 |
Seguridad y manipulación
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only