Learn More
3BP5L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA558233
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: CQQAEARVQA LQKTLRRAIG KSRPYFELKA QFSQILEEHK AKVTELEQQV AQAKTRYSVA LRNLEQISEQ IHARRRGGLP PHPLGPRRSS PVGAEAGPED Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
3BP5L Polyclonal antibody specifically detects 3BP5L in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
3BP5L | |
Polyclonal | |
Unconjugated | |
SH3BP5L | |
KIAA1720; SH3 binding domain protein 5 like; SH3 domain-binding protein 5-like; SH3-binding domain protein 5 like; SH3-binding domain protein 5-like; SH3BP5L; SH3BP-5-like; UNQ2766/PRO7133 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80851 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L8J4 | |
SH3BP5L | |
Recombinant protein corresponding to Human 3BP5L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only