Learn More
4-1BB Ligand Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA547291
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse 4-1BB Ligand, recombinant human (rh) APRIL, rhBAFF, rhEDA-A2, rhFas Ligand, rhGITR Ligand, rhLIGHT, rhOX40 Ligand, rhTNF-alpha, rhTRAIL, and rhTWEAK is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
4-1BB Ligand Polyclonal antibody specifically detects 4-1BB Ligand in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
4-1BB Ligand | |
Polyclonal | |
Unconjugated | |
TNFSF9 | |
4 1 BB Ligand; 4 1 BBL; 41BB Ligand; 4-1BB ligand; 41BBL; 4-1BBL; 4-1BB-L; AI848817; CD 137 Ligand; CD 137L; CD137 Ligand; CD137L; Cd157l; CDw137L; costimulatory molecule; homolog of mouse 4-1BB-L; Ly63l; receptor 4-1BB ligand; TNFRSF9; Tnfsf9; TNLG5A; tumor necrosis factor (ligand) superfamily member 9; tumor necrosis factor (ligand) superfamily, member 9; tumor necrosis factor ligand 5A; tumor necrosis factor ligand superfamily member 9; tumor necrosis factor superfamily member 9 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
8744 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P41273 | |
TNFSF9 | |
E. coli-derived recombinant human 4-1BB Ligand/TNFSF9 Arg71-Glu254 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |