Learn More
4,5-Diciano-2-(4-metilfenil)imidazol, 97 %, Thermo Scientific™
31.10€ - 391.00€
Identificadores de productos químicos
CAS | 63857-91-0 |
---|---|
Fórmula molecular | C12H8N4 |
Peso molecular (g/mol) | 208.224 |
Número MDL | MFCD20265230 |
Clave InChI | BMTQOJUWHGWRPD-UHFFFAOYSA-N |
Sinónimo | 4,5-dicyano-2-4-methylphenyl imidazole, 2-p-tolyl-1h-imidazole-4,5-dicarbonitrile, 2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-p-tolyl imidazole |
PubChem CID | 21798437 |
Nombre IUPAC | 2-(4-metilfenil)-1H-imidazol-4,5-dicarbonitrilo |
SMILES | CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
63857-91-0 | |
208.224 | |
BMTQOJUWHGWRPD-UHFFFAOYSA-N | |
21798437 | |
CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N |
C12H8N4 | |
MFCD20265230 | |
4,5-dicyano-2-4-methylphenyl imidazole, 2-p-tolyl-1h-imidazole-4,5-dicarbonitrile, 2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-p-tolyl imidazole | |
2-(4-metilfenil)-1H-imidazol-4,5-dicarbonitrilo |
Especificaciones
4,5-Dicyano-2-(4-methylphenyl)imidazole | |
C12H8N4 | |
4,5-dicyano-2-4-methylphenyl imidazole, 2-p-tolyl-1h-imidazole-4,5-dicarbonitrile, 2-4-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-p-tolyl imidazole | |
CC1=CC=C(C=C1)C2=NC(=C(N2)C#N)C#N | |
208.224 | |
208.22 |
63857-91-0 | |
MFCD20265230 | |
BMTQOJUWHGWRPD-UHFFFAOYSA-N | |
2-(4-metilfenil)-1H-imidazol-4,5-dicarbonitrilo | |
21798437 | |
97% |
Seguridad y manipulación
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes