Learn More
Sulfuro de 4-acetildifenilo, 97 %, Thermo Scientific™
Código de producto. 11400571
Click to view available options
Cantidad:
1 g
5 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
CAS 10169-55-8
Fórmula molecular C14H12OS
Peso molecular (g/mol) 228.31
Número MDL MFCD00026227
Clave InChI XUDYHODVSUXRPW-UHFFFAOYSA-N
Sinónimo 4-acetyldiphenyl sulfide, 1-4-phenylthio phenyl ethanone, 4-acetyl diphenyl sulfide, 4'-phenylthio acetophenone, 1-4-phenylsulfanyl phenyl ethanone, 1-4-phenylsulfanylphenyl ethanone, 1-4-phenylthio phenyl ethan-1-one, ethanone, 1-4-phenylthio phenyl, 1-acetyl-4-phenylthiobenzene
PubChem CID 66287
Nombre IUPAC 1-[4-(phenylsulfanyl)phenyl]ethan-1-one
SMILES CC(=O)C1=CC=C(SC2=CC=CC=C2)C=C1
Especificaciones
Nombre del producto químico o material 4-Acetyldiphenyl sulfide
Punto de fusión 64°C to 68°C
Punto de ebullición 183°C to 184°C (3 mmHg)
Olor Odorless
Cantidad 1 g
Beilstein 2049296
Formula Weight (peso de la fórmula) 228.32
Porcentaje de pureza 97%