missing translation for 'onlineSavingsMsg'
Learn More

4-Cloro-2-fluorotolueno, 99 %, Thermo Scientific™

Código de producto. 11354759
Click to view available options
Cantidad:
25 g
100 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 452-75-5
Fórmula molecular C7H6ClF
Peso molecular (g/mol) 144.57
Número MDL MFCD00000571
Clave InChI MKFCYQTVSDCXAQ-UHFFFAOYSA-N
Sinónimo 4-chloro-2-fluorotoluene, 2-fluoro-4-chlorotoluene, benzene, 4-chloro-2-fluoro-1-methyl, pubchem3437, acmc-209k1n, ksc493m8j, mkfcyqtvsdcxaq-uhfffaoysa, 4-chloro-2-fluoro-1-methyl-benzene
PubChem CID 520630
Nombre IUPAC 4-Cloro-2-fluoro-1-metilbenceno
SMILES CC1=CC=C(Cl)C=C1F

Specifications

Nombre del producto químico o material 4-Chloro-2-fluorotoluene
Densidad 1.184
Punto de ebullición 158°C to 160°C
Temperatura de inflamación 51°C (123°F)
Olor Aromatic
Índice de refracción 1.498
Cantidad 25 g
Número UN UN1993
Beilstein 1931682
Formula Weight (peso de la fórmula) 144.58
Porcentaje de pureza 99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.