Learn More
4-Etoxi-2-fluoroanilina, 97 %, Thermo Scientific™
Código de producto. 15466557
Click to view available options
Cantidad:
250 mg
1 g
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
CAS 470702-37-5
Fórmula molecular C8H10FNO
Peso molecular (g/mol) 155.17
Número MDL MFCD11053973
Clave InChI HDPGMJKNRMBTHT-UHFFFAOYSA-N
Sinónimo benzenamine, 4-ethoxy-2-fluoro, 4-ethoxy-2-fluorophenylamine, 4-ethoxy-2-fluoro-phenylamine, 4-ethoxy-2-fluoro-phenyl amine, benzenamine, 4-ethoxy-2-fluoro-9ci
PubChem CID 3734462
Nombre IUPAC 4-etoxi-2-fluoroanilina
SMILES CCOC1=CC(F)=C(N)C=C1
Especificaciones
Nombre del producto químico o material 4-Ethoxy-2-fluoroaniline
Cantidad 250 mg
Número UN UN2810
Formula Weight (peso de la fórmula) 155.17
Porcentaje de pureza 97%