Learn More
Hidrazida de ácido 4-fluorofenoxiacético, + 98 %, Thermo Scientific Chemicals
Código de producto. 11372926
Click to view available options
Cantidad:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS 1737-62-8
Fórmula molecular C8H9FN2O2
Peso molecular (g/mol) 184.17
Número MDL MFCD00082651
Clave InChI SEIHQPCBPNHCBN-UHFFFAOYSA-N
Sinónimo 2-4-fluorophenoxy acetohydrazide, 4-fluorophenoxyacetic acid hydrazide, 2-4-fluorophenoxy acetic acid hydrazide, acmc-1bpdj, p-fluoro-2-phenoxyacetohydrazide, 2-4-fluorophenoxy ;acetohydrazide, acetic acid,2-4-fluorophenoxy-, hydrazide
PubChem CID 735834
Nombre IUPAC 2-(4-fluorofenoxi)acetohidrazida
SMILES C1=CC(=CC=C1OCC(=O)NN)F
Especificaciones
Nombre del producto químico o material 4-Fluorophenoxyacetic acid hydrazide
Punto de fusión 124°C to 126°C
Cantidad 5 g
Beilstein 2526730
Formula Weight (peso de la fórmula) 184.17
Porcentaje de pureza ≥98%