Learn More
4E-BP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA561027
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: LLECKNSPIA RTPPCCLPQI PGVTTPPTAP LSKLEELKEQ ETEEEIPDDA QFEM Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 84%.
This gene encodes a member of the EIF4EBP family which derives it name from proteins that bind to eukaryotic initiation factor 4E and that prevent its assembly into EIF4F. Co-transcription of this gene and the neighboring upstream gene (MASK) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.Especificaciones
4E-BP3 | |
Polyclonal | |
Unconjugated | |
EIF4EBP3 | |
4EBP3; 4E-BP3; eIF4E-binding protein 3; EIF4EBP3; eukaryotic initiation factor 4E-binding protein 3; eukaryotic translation initiation factor 4E binding protein 3; eukaryotic translation initiation factor 4E-binding protein 3; NG36 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60516 | |
EIF4EBP3 | |
Recombinant protein corresponding to Human 4E-BP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only