missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4EBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA517745
Detalles adicionales : Peso : 0.01000kg
Descripción
It is not recommended to aliquot this antibody.
This gene encodes one member of a family of translation repressor proteins. The protein directly interacts with eukaryotic translation initiation factor 4E, which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.Especificaciones
4EBP1 | |
Polyclonal | |
Unconjugated | |
EIF4EBP1 | |
4EBP1; 4E-BP1; AA959816; BP-1; eIF4E-binding protein 1; EIF4EBP1; eukaryotic translation initiation factor 4E binding protein 1; eukaryotic translation initiation factor 4E-binding protein 1; MGC431; MGC4316; P/OKCL.6; PHAS-1; PHAS-I; Phosphorylated heat- and acid-stable protein regulated by insulin 1 | |
Rabbit | |
Protein A | |
RUO | |
116636, 13685, 1978 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
28.9 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
Q13541, Q60876, Q62622 | |
EIF4EBP1 | |
Synthetic peptide corresponding to residues surrounding Ser112 of human 4E-BP1 | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |