missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ácido 5-cloro-2-(trifluorometoxi)benzoico, 97 %, Thermo Scientific™
83.48€ - 331.00€
Identificadores de productos químicos
CAS | 959749-82-7 |
---|---|
Fórmula molecular | C8H4ClF3O3 |
Peso molecular (g/mol) | 240.56 |
Número MDL | MFCD11519354 |
Clave InChI | MXQBGMLCAJITNV-UHFFFAOYSA-N |
Sinónimo | 5-chloro-2-trifluoromethoxy benzoic acid, 5-chloro-2-trifluoromethoxy benzoic acid, jrd |
PubChem CID | 57361682 |
Nombre IUPAC | ácido 5-cloro-2-(trifluorometoxi)benzoico |
SMILES | OC(=O)C1=C(OC(F)(F)F)C=CC(Cl)=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
959749-82-7 | |
240.56 | |
MXQBGMLCAJITNV-UHFFFAOYSA-N | |
57361682 | |
OC(=O)C1=C(OC(F)(F)F)C=CC(Cl)=C1 |
C8H4ClF3O3 | |
MFCD11519354 | |
5-chloro-2-trifluoromethoxy benzoic acid, 5-chloro-2-trifluoromethoxy benzoic acid, jrd | |
ácido 5-cloro-2-(trifluorometoxi)benzoico |
Specifications
5-Chloro-2-(trifluoromethoxy)benzoic acid | |
83°C to 85°C | |
MFCD11519354 | |
MXQBGMLCAJITNV-UHFFFAOYSA-N | |
ácido 5-cloro-2-(trifluorometoxi)benzoico | |
57361682 | |
97% |
959749-82-7 | |
C8H4ClF3O3 | |
5-chloro-2-trifluoromethoxy benzoic acid, 5-chloro-2-trifluoromethoxy benzoic acid, jrd | |
OC(=O)C1=C(OC(F)(F)F)C=CC(Cl)=C1 | |
240.56 | |
240.57 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes