Learn More
53BP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554565
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: VPSPATRSEA LSSVLDQEEA MEIKEHHPEE GSSGSEVEEI PETPCESQGE ELKEENMESV PLHLSLTETQ SQGLCLQKEM PKKECSEAME VETSVISIDS PQKLA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
53BP1 (P53-binding protein 1) plays a critical role in tumor suppression and is a putative substrate of ATM kinase. Upon DNA damage, 53BP1 is phosphorylated and localizes to the presumptive sites of damage, specifically, double-strand breaks. 53BP1 may have a role in checkpoint signaling during mitosis, enhances TP53-mediated transcriptional activation, and participates in DNA repair by maintaining genomic stability.Especificaciones
53BP1 | |
Polyclonal | |
Unconjugated | |
TP53BP1 | |
53BP1; FLJ41424; m53BP1; MGC138366; murine p53-binding protein; p202; p53-binding protein 1; p53BP1; TDRD30; TP53; TP53-binding protein 1; Tp53bp1; transformation related protein 53 binding protein 1; transformation related protein 53 binding-protein 1; Trp53bp1; tumor protein 53-binding protein, 1; tumor protein p53 binding protein 1; tumor protein p53-binding protein, 1; tumor suppressor p53-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27223, 296099, 7158 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P70399, Q12888 | |
TP53BP1, Trp53bp1 | |
Recombinant protein corresponding to Human 53BP1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only