missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5T4 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Marca: Invitrogen PA547690
Detalles adicionales : Peso : 0.00010kg
Descripción
In direct ELISAs, approximately 5% cross-reactivity with recombinant human 5T4 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
5T4 Polyclonal antibody specifically detects 5T4 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Especificaciones
5T4 | |
Polyclonal | |
Unconjugated | |
TPBG | |
5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
21983 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9Z0L0 | |
TPBG | |
Mouse myeloma cell line NS0-derived recombinant mouse 5T4 Ser30-Ser361 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |