missing translation for 'onlineSavingsMsg'
Learn More

Ácido 9-fluorenona-4-carboxílico, +98 %, Thermo Scientific Chemicals

Catalog Number 11465977
Click to view available options
Cantidad:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
6223-83-2
C14H8O3
224.215
MFCD00001145
AFQYQSWTVCNJQT-UHFFFAOYSA-N
9-fluorenone-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid, 9h-fluorene-4-carboxylic acid, 9-oxo, 9-fluorenone-4-carboxlic acid, 9-oxofluorene-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid #
80361
Ácido 9-oxofluoreno-4-carboxílico
C1=CC=C2C(=C1)C3=C(C2=O)C=CC=C3C(=O)O
Este artículo no se puede devolver. Vea la política de devoluciones
6223-83-2
C14H8O3
224.215
MFCD00001145
AFQYQSWTVCNJQT-UHFFFAOYSA-N
9-fluorenone-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid, 9h-fluorene-4-carboxylic acid, 9-oxo, 9-fluorenone-4-carboxlic acid, 9-oxofluorene-4-carboxylic acid, 9-oxo-9h-fluorene-4-carboxylic acid #
80361
Ácido 9-oxofluoreno-4-carboxílico
C1=CC=C2C(=C1)C3=C(C2=O)C=CC=C3C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material 9-Fluorenone-4-carboxylic acid
Punto de fusión 224°C to 228°C
Cantidad 25 g
Beilstein 2694801
Formula Weight (peso de la fórmula) 224.22
Porcentaje de pureza ≥98%

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'