missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ 96 Deep Well Plate
Deep Well Plates for assays, sample storage, and mixing.
Marca: Cole-Parmer™ 16344-91
Detalles adicionales : Peso : 4.72700kg
Descripción
- Certified free of RNase, DNase, endotoxins, and human DNA
- Notched corners to facilitate orientation
- Compatible with most automated liquid handling systems
- Can be autoclaved at 121°C for 20 minutes
- Raised rims provide efficient sealing to minimize the risk of cross-contamination
- Round U-shaped wells reduce sample loss while pipetting
- Available gamma sterilized
- Use with sealing films and pads (order separately in table) to lessen sample evaporation and contamination
Especificaciones
No Barcode | |
8 x 12 | |
Polypropylene | |
96 | |
Non-treated | |
50 |
Standard | |
Clear | |
1.6 mL | |
Sterile | |
Round Bottom (U-well) |