missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ABL1 [F317L], GST Tag Recombinant Protein

This ABL1 F317L Recombinant Human Protein (27-end) was expressed in insect cells.

Marca:  Invitrogen™ A33550

 Ver más versiones de este producto

Código de producto. 17198805

  • 2435.00€ / 100 microgramos

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

ABL1 cytoplasmic tyrosine kinase protooncogene encodes a cytoplasmic and nuclear kinase that has been implicated in processes of cell differentiation, cell division, cell adhesion, and stress response. Activity of ABL protein is negatively regulated by its SH3 domain and deletion of the SH3 domain turns ABL1 into an oncogene. Translocation and head-to-tail fusion of the BCR and ABL1 genes is present in many cases of chronic myelogeneous leukemia. The DNA-binding activity of the ubiquitously expressed ABL1 tyrosine kinase is regulated by CDK1-mediated phosphorylation, suggesting a cell cycle function for ABL1.
  • Validated for use with the Z’-LYTE™ assay
  • Available in multiple pack sizes for convenient use
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

NP_005148.2
146.3 kDa
ABL Kinase Family
Human
GST Tag
Dry Ice
Recombinant, Active, Mutant
Kinase Assay
100 μg
aa 27 - end
Recombinant
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.
Insect
Liquid
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales