Learn More
Abnova™ Kit de ensayo de L-alanina
Se utiliza para detectar alanina y el estudio funcional.
Includes:
  • Alanine assay buffer, 25mL
  • Alanine probe (in DMSO), 0.2mL
  • Alanine converting enzyme (lyophilized), 1 vial
  • Alanine development mix (lyophilized), 1 vial
  • Alanine standard (10μmol, lyophilized), 1 vial