missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic Anhydride, Merck Chemicals™
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Click to view available options
Cantidad:
1L
Especificaciones
CAS | 108-24-7 |
Densidad | 1.08g/cm3 |
Fórmula molecular | C4H6O3 |
Cantidad | 1L |
Sinónimo | acetic anhydride, acetanhydride, acetyl oxide, acetic acid, anhydride, acetic oxide, acetyl anhydride, anhydride acetique, ethanoic anhydride, acetyl ether, acetic acid anhydride |
Clave InChI | WFDIJRYMOXRFFG-UHFFFAOYSA-N |
Peso molecular (g/mol) | 102.09 |
PubChem CID | 7918 |
ChEBI | CHEBI:36610 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'