Learn More
ACROBiosystems ActiveMax™ Human VEGF110 Protein, Tag Free
HPLC-verified
Marca: ACROBiosystems VE0-H5212-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Vascular endothelial growth factor (VEGF), also known as vascular permeability factor (VPF) and VEGF-A, and is a member of the platelet-derived growth factor (PDGF)/vascular endothelial growth factor (VEGF) family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. Alternatively spliced isoforms of 110,121,145,165,183,189 and 206 amino acids in length are expressed in humans.
This protein carries no "e;tag"e;. The protein has a calculated MW of 12.7 kDa. The protein migrates as 16-18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_001165097 | |
200 μg | |
Ala 27 - Arg 136 | |
RP1-261G23.1,MGC70609,MVCD1,VEGFA,VPF | |
Unconjugated | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
12.7 kDa | |
ActiveMax™ Human VEGF110, Tag Free (HPLC-verified) (VE0-H5212) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 136 (Accession # NP_001165097). | |
-20°C | |
VEGF110 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
ActiveMax™ Cytokines |