missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine Deaminase Monoclonal Antibody (CL0176), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524586
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: SDNQPEGMIS ESLDNLESMM PNKVRKIGEL VRYLNTNPVG GLLEYARSHG FAAEFKLVDQ SGPPHEPKFV YQAKVGGRWF PAVCAHSKKQ GKQEAADAAL RVLIGENEKA ERMGFTEVTP VTGASLRRTM LLLSRSPEA Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
Catalyzes the hydrolytic deamination of adenosine and 2-deoxyadenosine. Plays an important role in purine metabolism and in adenosine homeostasis. Modulates signaling by extracellular adenosine, and so contributes indirectly to cellular signaling events. May act as a positive regulator of T-cell coactivation.Especificaciones
Adenosine Deaminase | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P00813 | |
ADA | |
Recombinant protein corresponding to Human Adenosine Deaminase | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
CL0176 | |
Unconjugated | |
ADA | |
ada; ADA1; adenosine aminohydrolase; Adenosine deaminase; RP11-61L14.5; zgc:92028 | |
Mouse | |
Protein A | |
RUO | |
100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido