missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Kinase 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA564107
350.00 EUR hasta el 2025-03-16
Use el código promocional "24070" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: GKGTVSSRIT THFELKHLSS GDLLRDNMLR GTEIGVLAKA FIDQGKLIPD DVMTRLALHE LKNLTQYSWL LD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 93%.
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1.
Especificaciones
Adenylate Kinase 3 | |
Polyclonal | |
Unconjugated | |
AK3 | |
AA407498; adenylate kinase 3; adenylate kinase 3 {ECO:0000255; Adenylate kinase 3 alpha-like 1; adenylate kinase 3 alpha-like 1 {ECO:0000255; adenylate kinase 6, adenylate kinase 3 like 1; AI506714; AK 3; AK 3 {ECO:0000255; AK3; AK-3; Ak3l; AK3L1; AK4; AK6; AKL3L; AKL3L1; FIX; GTP:AMP phosphotransferase AK3, mitochondrial; GTP:AMP phosphotransferase AK3, mitochondrial {ECO:0000255; GTP:AMP phosphotransferase mitochondrial; GTP:AMP phosphotransferase, mitochondrial; HAMAP-Rule:MF_03169} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UIJ7 | |
AK3 | |
Recombinant protein corresponding to Human Adenylate Kinase 3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Adenylate Kinase 3 Polyclonal Antibody, Invitrogen™ > 100 μL; Unconjugated
Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido