missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK5 Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501641S

This item is not returnable.
View return policy
Description
AK5 Monoclonal antibody specifically detects AK5 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
AK5 | |
Monoclonal | |
0.91 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9Y6K8 | |
Mouse | |
Affinity chromatography | |
RUO | |
26289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G4 | |
Unconjugated | |
AK5 | |
adenylate kinase 5; adenylate kinase 6; adenylate kinase isoenzyme 5; AK 5; AK5; AK6; ATP-AMP transphosphorylase 5; MGC33326; wu:fj63a06 | |
Full length human recombinant protein of human AK5 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AK5 Monoclonal Antibody (OTI2G4), TrueMAB™, OriGene
Spot an opportunity for improvement?Share a Content Correction