missing translation for 'onlineSavingsMsg'
Learn More

ALDH5A1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA529096

Código de producto. 13473217

  • 514.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse brain, A549. Predicted reactivity: Mouse (84%), Rat (84%), Pig (85%), Chimpanzee (92%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Aldh5A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldehyde dehydrogenase enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. Aldh5A1 is a mitochondrial NAD(+)-dependent succinic semialdehyde dehydrogenase. A deficiency of this enzyme, known as 4-hydroxybutyricaciduria, results in a disorder of the neurotransmitter 4-aminobutyric acid (GABA). Symptoms usually include static encephalopathy, associated with developmental delays, hypotonia, ataxia, speech defects, and seizures. At least two isoforms of Aldh5A1 are known to exist.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

ALDH5A1
Polyclonal
Unconjugated
ALDH5A1
6330403E24Rik; Ahd1; Ahd-1; aldehyde dehydrogenase 1, mitochondrial; aldehyde dehydrogenase 5 family member A1; aldehyde dehydrogenase 5 family, member A1; aldehyde dehydrogenase 5 family, member A1 (succinate-semialdehyde dehydrogenase); aldehyde dehydrogenase family 5 member A1; aldehyde dehydrogenase family 5, subfamily A1; ALDH5A1; aldhehyde dehydrogenase family 5, subfamily A1; D630032B01Rik; mitochondrial succinate semialdehyde dehydrogenase; NAD(+)-dependent succinic semialdehyde dehydrogenase; OTTMUSG00000000613; SSADH; SSDH; Ssdh1; Succinate-semialdehyde dehydrogenase, mitochondrial; succinic semialdehyde dehydrogenase
Rabbit
Antigen affinity chromatography
RUO
214579, 7915
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P51649, Q8BWF0
ALDH5A1
Recombinant protein fragment corresponding to a region within amino acids 85 and 324 of Human ALDH5A1
100 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales