missing translation for 'onlineSavingsMsg'
Learn More

alpha Actinin 3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA527862

Código de producto. 13411827

  • 473.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Recommended positive controls: 293T, A431, HeLa, HepG2, NIH-3T3, JC, PC-12. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (94%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Alpha-actinin-3, also known as alpha-actinin skeletal muscle isoform 3 or F-actin cross-linking protein, is a protein that in humans is encoded by the ACTN3 gene. This gene encodes a member of the alpha-actin binding protein gene family. The encoded protein is primarily expressed in skeletal muscle and functions as a structural component of sarcomeric Z line. This protein is involved in crosslinking actin containing thin filaments. An allelic polymorphism in this gene results in both coding and non-coding variants; the reference genome represents the coding allele.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

alpha Actinin 3
Polyclonal
Unconjugated
Actn3
actinin alpha 3; actinin alpha 3 (gene/pseudogene); Actinin alpha3; actinin, alpha 3; Actinin-alpha3; ACTN3; alpha-actinin skeletal muscle; Alpha-actinin skeletal muscle isoform 3; alpha-Actinin3; Alpha-actinin-3; alphaActinin-3; F-actin cross-linking protein
Rabbit
Antigen affinity chromatography
RUO
11474, 171009, 89
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
O88990, Q08043
Actn3
Recombinant fragment corresponding to a region within amino acids 1 and 363 of Human alpha Actinin 3
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales