missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Internexin Monoclonal Antibody (2E3), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen 323600
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is purified from mouse ascites fluid and is specific for the 66 kDa alpha-internexin protein. 2E3 antibody has stained human neuron and nerve fiber in human cerebral cortex. It has also stained glioma, ganglioneuroma, pheochromocytoma, and paraganglioma in human tissue. It is recommended that FFPE tissues are pretreated with HIER using citrate buffer, pH 6.
alpha Internexin Monoclonal antibody specifically detects alpha Internexin in Human, Mouse, Rabbit, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot.Especificaciones
alpha Internexin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P23565, P46660, Q16352 | |
INA | |
Full-length recombinant rat alpha-internexin protein. | |
100 μg | |
Primary | |
Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
2E3 | |
Unconjugated | |
INA | |
66 kDa neurofilament protein; Alpha-internexin; alpha-Inx; AV028420; FLJ18662; FLJ57501; Ina; Inexa; internexin neuronal intermediate filament protein alpha; internexin neuronal intermediate filament protein, alpha; internexin, alpha; Intlaa; LOW QUALITY PROTEIN: alpha-internexin; MGC12702; NEF5; Neuro; Neurofilament 5; neurofilament 5 (66kD); Neurofilament 66; neurofilament-66; neurofilament-66, tax-binding protein; Nf66; NF-66; TXBP-1 | |
Mouse | |
Protein A | |
RUO | |
100348512, 226180, 24503, 9118 | |
-20°C | |
Liquid |