Learn More
STEMCELL Technologies Alpha MEM without Nucleosides
Minimum Essential Medium Eagle - alpha modification (Alpha MEM) without nucleosides
Marca:  STEMCELL Technologies 36453
Encompass
Código de producto. 17168091
  • 88.50€ / 500 ml
Explore más ofertas especiales
Descripción
Descripción
  • Minimum Essential Medium Eagle - Alpha Modification (Alpha MEM) without Nucleosides is recommended for a wide variety of cell culture applications
  • Selection of a suitable nutrient medium is dependent on type of cell, conditions of culture, and degree of chemical definition
  • MEM has typically been used for the cultivation of cells grown in monolayers; however, additional supplementation to Alpha MEM has expanded the applications for this medium
Especificaciones
Especificaciones
500 mL