Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
Click to view available options
Tamaño:
Small
XL
2XL
Descripción
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
Especificaciones
Tamaño | Small |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Tipo de costura | Bound |
Certificaciones/conformidad (Europa) | Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 |
Modelo | W20B301 |
Para utilizar con (aplicación) | Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |