Learn More
Ansell AlphaTec™ 2000 STANDARD Trousers Bound
Catalog Number 16347686
Click to view available options
Tamaño:
Small
XL
2XL
Comfortable trousers featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress.
  • Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
  • Optimized fit, improving wearer comfort and safety
  • Low Linting
Tamaño Small
Material Microporous Non-woven Polyethylene Laminate
Color White
Tipo de costura Bound
Certificaciones/conformidad (Europa) Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013
Modelo W20B301
Para utilizar con (aplicación) Clean Room Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services