Learn More
Ansell Alphatec™ 3000 Apron with Sleeves Ultrasonically Welded - Model 243-G01
Click to view available options
Tamaño:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
DESIGN FEATURES:
- Apron with sleeves with rear velcro fastening at the neck and attached AlphaTec™ 37-676
- Unexpected leakages spills or other releases
- Loading and unloading trucks and vehicles
- Loading trucks and vehicles
- Product inspection
- Cleaning of plant and machines
Specifications
Specifications
Tipo | Chemical-resistant Apron |
Tamaño | Small |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Desechables | Reusable |
Tipo de tapón | Hook and Loop |
Esterilidad | Non-sterile |
Peso (métrico) | 80 gsm |
Tipo de costura | Welded |
Certificaciones/conformidad (Europa) | CE CATEGORY III (Type PB [3]-B, Type PB [4]-B, EN 14126) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction