Learn More
Ansell™ AlphaTec™ 4000 Ultrasonically Welded and Taped
963.18 EUR hasta el 2024-12-31
Use el código promocional "22917" para aplicar la oferta.
Use el código promocional "22917" para aplicar la oferta.
Click to view available options
Tamaño:
Small
Medium
Large
2X-Large
3X-Large
Descripción
Consists of a unique multi-layer barrier fabric that provides lightweight and robust textile feel with exceptional barrier to organic and inorganic chemicals.
- Exceptional protection, over 200 chemicals permeation tested, including chemical warfare agents
- Designed to protect, typical innovative features include dual zip systems and double cuffs
- Welded and taped seams, our highest barrier to liquids and particulates
- Enhanced comfort, knitted inner cuffs and inner textile type fabric improves wearer acceptance
- Anti-static, tested according to EN 1149-5
Especificaciones
Tipo de producto | Chemical Protective Clothing |
Tamaño | Small |
Sexo | Unisex |
Material | Multi-layer Barrier Laminate Fabric |
Color | Spruce Green |
Desechables | Reusable |
Estilo de cintura | Elastic |
Estilo de tobillo | Elastic |
Estilo de manguito | Elastic, Double Inner Knit |
Estilo de cuello | Hooded |