Learn More
Varilla de nitruro de aluminio, redonda, 22 mm de diámetro, Alfa Aesar™
Click to view available options
Cantidad:
150 mm
200 mm
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
Nombre del producto químico o material | Aluminum nitride |
Color | Blanco a amarillo |
Aspecto | White to pale yellow rod |
Cantidad | 200 mm |
Nombre IUPAC | azanilidinoalumano |
Olor | Odorless |
Formulario | Varilla |