missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMER1 Monoclonal Antibody (729419), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524343
Detalles adicionales : Peso : 0.00010kg
Descripción
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
AMER1 Monoclonal antibody specifically detects AMER1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
AMER1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q5JTC6 | |
AMER1 | |
E. coli-derived recombinant human WTX Met421-Leu573 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunocytochemistry, Western Blot | |
729419 | |
Unconjugated | |
AMER1 | |
2810002O09Rik; adenomatous polyposis coli membrane recruitment 1; Amer1; APC membrane recruitment 1; APC membrane recruitment protein 1; AW492303; Fam123b; family with sequence similarity 123, member B; family with sequence similarity 123B; OSCS; protein Fam123b; RGD1560322; RP11-403E24.2; Wilms tumor gene on the X chromosome protein; Wilms tumor on the X; WTX | |
Mouse | |
Protein A/G | |
RUO | |
139285 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |