missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amoxicillin sodium salt, 89% min., Thermo Scientific Chemicals
Click to view available options
Cantidad:
5 g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Nombre del producto químico o material | Amoxicillin sodium salt |
Tipo | Amoxicillin Sodium Salt |
Forma física | Powder |
Punto de fusión | 130°C |
Información de solubilidad | Soluble in water. |
Formula Weight (peso de la fórmula) | 387.39 |
Porcentaje de pureza | ≥89% |
Color | Blanco |
Cantidad | 5 g |
Temperatura de inflamación | >110°C (230°F) |
Afficher plus |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'