Learn More
AMPK alpha-1 Recombinant Rabbit Monoclonal Antibody (SU03-48), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Marca: Invitrogen MA532122
Descripción
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains.Especificaciones
AMPK alpha-1 | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide, pH 7.4 | |
P54645, Q13131, Q5EG47 | |
Prkaa1 | |
Synthetic peptide within Human AMPK alpha aa 501-550 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Western Blot, Western Blot | |
SU03-48 | |
Unconjugated | |
Prkaa1 | |
5'-AMP-activated protein kinase alpha-1 catalytic subunit, 5'-AMP-activated protein kinase catalytic subunit alpha-1, 5'-AMP-activated protein kinase, catalytic alpha-1 chain, 63 kDa subunit, AAPK2, ACACA kinase, Acetyl-CoA carboxylase kinase, activated protein kinase, AI194361, AI450832, AL024255, AMP -activate kinase alpha 1 subunit, AMP activated protein kinase, AMP-activated protein kinase, alpha 1 catalytic subunit, AMP-activated protein kinase, catalytic, alpha-1, AMPK, AMPK 63 kDa subunit, AMPK alpha 1, AMPK alpha-1 chain, AMPK subunit alpha-1, AMPK subunit pAMPK, AMPK1, AMPK2, AMPKa1, AMPKalpha1, AMPK-alpha-2 chain, C130083N04Rik, HMGCR kinase, hydroxymethylglutaryl-CoA reductase kinase, MGC33776, MGC57364, pAMPK, PRKAA, Prkaa1, PRKAA2, protein kinase, protein kinase AMP-activated catalytic subunit alpha 1, protein kinase, AMP-activated, alpha 1 catalytic subunit, tau-protein kinase PRKAA1 | |
Rabbit | |
Protein A | |
RUO | |
105787, 5562, 65248 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.