missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Angiotensin II Antibody (KAA8), Alexa Fluor™ 488 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-20123AF488
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Angiotensin II Monoclonal antibody specifically detects Angiotensin II in Human, Rat samples. It is validated for Western BlotEspecificaciones
Angiotensin II | |
Monoclonal | |
Alexa Fluor 488 | |
AGT, alpha-1 antiproteinase, antitrypsin, angiotensin I, ANHU, pre-angiotensinogen, serine (or cysteine) proteinase inhibitor, Serpin A8 | |
Angiotensin II | |
0.1 mL | |
Primary | |
Human, Rat | |
Purified |
Western Blot | |
KAA8 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
183 | |
Store at 4°C in the dark. | |
IgG κ |