Learn More
anti-alpha Amylase, Polyclonal PA129937, Invitrogen™
Specifically detect alpha Amylase in Bacteria samples using Thermo Scientifc™ Pierce™ anti-alpha Amylase Polyclonal, validated for ELISA, IP and WB
Marca: Thermo Scientific PA1-29937
Descripción
Specifically detect alpha Amylase in Bacteria samples using Thermo Scientifc™ Pierce™ anti-alpha Amylase Polyclonal, validated for ELISA, IP and WB
Host Species: Rabbit
Cross Reactivity: Bacteria
Isotype: IgG
Immunogen: Alpha Amylase [Bacillus amyloliquefaciens]
Formulation: 1mL of Unpurified antibody in whole serum, with NaCl 0.15 M and potassium phosphate 0.02 M diluted in potassium phosphate 0.02 M; pH 7.2.
PA1-29937 detects alpha Amylase in Bacteria samples. PA1-29937 has been successfully used in ELISA, IP and WB procedures.PA1-29937 The immunogen is Alpha Amylase [Bacillus amyloliquefaciens].
Especificaciones
| alpha Amylase | |
| Polyclonal | |
| Rabbit | |
| 1 mL | |
| Primary | |
| Store at 4°C. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Alpha Amylase [Bacillus amyloliquefaciens] | |
| RUO | |
| Bacteria | |
| Purified |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.