Learn More
BAF47 Mouse, Unlabeled, Clone: 25, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 612110
Detalles adicionales : Peso : 0.01000kg
Descripción
SWI/SNF complexes facilitate gene activation and transcription factor binding by altering repressive chromatin structures in an ATP-dependent manner. In mammals, SWI/SNF complexes are present in mutliple forms that include 9-12 BRG1-associated factors (BAFs) per complex. These BAF proteins range in molecular weight from 47 to 250kDa. BAF47/SNF5 (SMARCB1/Ini1) complexes with BRG1- and BRM-containing SWI/SNF complexes. BAF47/SNF5 mRNA is widely expressed in cell lines and tissues, and BAF47/SNF5 gene is mutated in many human tumors. The latter is indicative of a tumor suppressor role for BAF47/SNF5. In addition, mice deficient for BAF47 die early in embryogenesis, while BAF47/SNF5 heterozygous mice display a variety of tumors in the soft tissues of the head and neck. BAF47/SNF5 also binds the HIV-1 integrase and stimulates integrase-mediated DNA joining activity. Thus, BAF47/SNF5 is a component of SWI/SNF complexes that may be critical for normal development and tumor suppression, but may also be a protein utilized for viral DNA integration into host DNA.
Western Blotting
Especificaciones
BAF47 | |
Monoclonal | |
250μg/mL | |
SNF5 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
25 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Mouse BAF47 aa. 257-359 | |
50 μg | |
Cell Biology | |
Human, Mouse, Rat | |
IgG2a |
For Research Use Only.