Learn More
c-Raf Mouse, Unlabeled, Clone: 53, BD
Mouse Monoclonal Antibody
374.00€ - 595.00€
Especificaciones
Antígeno | c-Raf |
---|---|
Clon | 53 |
Concentración | 250μg/mL |
Aplicaciones | Western Blot |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15895289
|
BD Biosciences
610151 |
50 μg |
374.00€
1 set |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15805299
|
BD Biosciences
610152 |
150 μg |
595.00€
1 set |
Fecha estimada de envío: 13-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Raf (c-Raf-1) is a cytoplasmic serine/threonine protein kinase and is a member of a family of proteins which are highly conserved from Drosophila to mammals. Raf has a critical role in the response to many growth factors including: EGF, PDGF, insulin, IL-2, IL-3, CSF-1, and GM-CSF. Raf can directly interact with Ras-GTP and subsequently become activated. Thus Raf plays a prominent role in the Ras signaling pathway by transferring a signal from Ras which has been activated by growth factor receptor-stimulated tyrosine kinase activity. This leads to the stimulation and activation of a number of other cytoplasmic serine/threonine kinases. Raf regulates the MAP kinase pathway by phosphorylating and activating MEK, which then phosphorylates and activates MAP kinase (ERK). ERK then phosphorylates and activates Rsk. Raf activity can also be regulated independently of Ras. Cyclic AMP (cAMP) activation of protein kinase A (PKA) can inhibit growth factor stimulation of Raf.
Especificaciones
c-Raf | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
53 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.