missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CCR7, PE, Clone: 4B12, R&D Systems™
Rat Monoclonal Antibody has been used in 4 publications
Marca: R&D Systems FAB3477P-100
Detalles adicionales : Peso : 0.00970kg
Descripción
CCR7 Monoclonal specifically detects CCR7 in Mouse samples. It is validated for Flow Cytometry.Especificaciones
CCR7 | |
Monoclonal | |
R-PE | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
BLR2, BLR2C-C chemokine receptor type 7, C-C CKR-7, CC-CKR-7, CCR7, CCR-7, CD197, CD197 antigen, CDw197, CDw197CC chemokine receptor 7, chemokine (C-C motif) receptor 7, CMKBR7, CMKBR7chemokine (C-C) receptor 7, EBI1, EBI1EBV-induced G protein-coupled receptor 1, EBV-induced G-protein coupled receptor 1, Epstein-Barr virus induced gene 1, Epstein-Barr virus induced G-protein coupled receptor, Epstein-Barr virus-induced G-protein coupled receptor 1, EVI1, lymphocyte-specific G protein-coupled peptide receptor, MIP-3 beta receptor | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects mouse CCR7. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG2a |
Flow Cytometry | |
4B12 | |
Flow Cytometry 10 uL/10^6 cells | |
NP_031745 | |
CCR7 | |
RBC-2H3 cells expressing mouse CCR7 Accession # NP_031745 | |
100 Tests | |
1236 | |
Mouse | |
Purified |