Learn More
CD112 Mouse anti-Human, Unlabeled, Clone: R2.525, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 551056
146.66 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.
Descripción
Reacts with the poliovirus receptor 2 (PRR2) protein. PRR2 is a cell surface molecule homologous to the poliovirus receptor (PVR/CD155). It is a transmembrane glycoprotein belonging to the immunoglobulin superfamily. The PRR2 gene encodes for two mRNA isoforms, a short form of 3.0 kb (hPRR2α) and a long form of 4.4 kb (hPRR2δ). Both are widely expressed in human tissues, including hematopoietic cells. PRR2 is also expressed on endothelial cells, at the intercellular junctions of adjacent cells. It has been suggested that PRR2 may play a role in the regulation of hematopoietic/endothelial cell interaction.

Especificaciones
CD112 | |
Monoclonal | |
0.5mg/mL | |
PRR2 | |
Mouse | |
0.1 mg | |
Cell Biology | |
Human | |
IgG1 κ |
Flow Cytometry | |
R2.525 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.