Learn More
CD114 Mouse anti-Human, PE, Clone: LMM741, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 554538
Descripción
The LMM741 antibody reacts with the 150 kD human granulocyte-colony stimulating factor receptor (G-CSFR) also known as CD114. Cell surface G-CSFR is expressed by neutrophils, myeloid leukemias, endothelium, platelets, and placenta. The immunogen used to generate this hybridoma was cells transfected with an expression vector containing a full-length cDNA encoding the G-CSF-receptor.
Especificaciones
| CD114 | |
| Monoclonal | |
| 0.2mg/mL | |
| G-CSF Receptor | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| LMM741 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| G-CSFR cDNA transfected cells. | |
| 0.2 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.